MilliporeSigma™ LiChrospher™ 100 RP-8 Sorbents
well suited for the separation of neutral, acidic and weak basic compounds.
$2,723.54 - $2,934.80
Specifications
Material Silica
Type HPLC Sorbent
Quantity 10 g
Surface Area (Metric) 350 m2/g
View More Specs
Products 2
Catalog Number Mfr. No. Particle Size Price Quantity  
Catalog Number Mfr. No. Particle Size Price Quantity  
M1161290010
View Documents
MilliporeSigma™
1161290010
5 µm
Each for $2,934.80
M1161390010
View Documents
MilliporeSigma™
1161390010
10 µm
Each for $2,723.54
Description
Description
LiChrospher™ RP-8 is a reliable and versatile traditionally produced spherical silica gel carrier with C-8 reversed phase properties.
Specifications
Specifications
Silica
HPLC Sorbent
10 g
350 m2/g