Please login to your online account to display your discounted pricing
EMD Millipore LiChrolut™ Sorbents
Constant batch-to-batch quality ensured with quality control of raw materials
Manufacturer: emd millipore  1196860001
 View more versions of this product
Catalog No. M1196860001
  • / Each
Description & Specifications
Specifications
Product Type SPE Sorbent
Surface Area (Metric) approx.600m2/g
Sorbent LiChrolut RP-18
Particle Size 40 to 63μm
Material Polypropylene (tube)
Quantity 30/Pk.