missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ LiChroCART™ Columns/Purospher™ STAR Si Sorbents HPLC Cartridges, 5μm
Offers exceptional separation efficiency for normal-phase chromatography of low molecular weight compounds soluble in organic solvents
$2,042.29 - $2,159.03
Specifications
Column Format | Cartridge |
---|---|
Max. Pressure | 2175 psi (150 bar) to 2900 psi (200 bar) |
Particle Size | 5 μm |
pH | 2 to 7.5 |
Pore Size | 120 Å |
Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Price | Quantity | ||||
M1502680001
|
MilliporeSigma™
1502680001 |
4 mm | 125 mm |
Each for $2,042.29
|
|
M1502690001
|
MilliporeSigma™
1502690001 |
4 mm | 250 mm |
Each for $2,159.03
|
|
M1503560001
|
MilliporeSigma™
1503560001 |
4.6 mm | 150 mm |
Each for $2,135.71
|
|
M1503570001
|
MilliporeSigma™
1503570001 |
4.6 mm | 250 mm |
Each for $2,159.03
|
|
Description
- Very high separation efficiency as measured by the plate count
- Absence of metal impurities, thus giving consistently symmetrical peaks
- Extended column lifetime
Specifications
Cartridge | |
5 μm | |
120 Å | |
330 m²/g | |
Silica | |
Normal Phase | |
HPLC Cartridge |
2175 psi (150 bar) to 2900 psi (200 bar) | |
2 to 7.5 | |
1/Pk. | |
L3 | |
Silica | |
Normal Phase |