Please login to your online account to display your discounted pricing

EMD Millipore LiChroCART™ Columns/Purospher™ STAR Si Sorbents HPLC Cartridges, 5μm


Offers exceptional separation efficiency for normal-phase chromatography of low molecular weight compounds soluble in organic solvents

Manufacturer: emd millipore  1502690001

 View more versions of this product

Catalog No. M1502690001

  • / Each
 

Description & Specifications

Specifications

Diameter (English) Inside 4mm
Type HPLC Cartridges
Length (Metric) 250mm
Quantity 1/Pk.