Please login to your online account to display your discounted pricing

EMD Millipore LiChroCART™ Columns/Purospher™ STAR NH2 Sorbents HPLC Cartridges, 5μm


Designed for carbohydrate analysis with a typical mobile phase consisting of acetonitrile and water

Manufacturer: emd millipore  1502480001

 View more versions of this product

Catalog No. M1502480001

  • / Each
 

Description & Specifications

Specifications

Quantity 1/Pk.
Length (Metric) 250mm
Type HPLC Cartridges
Diameter (English) Inside 4.6mm