Please login to your online account to display your discounted pricing

EMD Millipore LiChroCART™ Columns/Purospher™ STAR NH2 Sorbents HPLC Cartridges, 5μm


Designed for carbohydrate analysis with a typical mobile phase consisting of acetonitrile and water

Manufacturer: emd millipore  1502450001

 View more versions of this product

Catalog No. M1502450001

  • / Each
 

Description & Specifications

Specifications

Type HPLC Cartridges
Length (Metric) 250mm
Quantity 1/Pk.
Diameter (English) Inside 4mm