Please login to your online account to display your discounted pricing
EMD Millipore LiChroCART™ Column/Superspher™ Sorbent HPLC Cartridges, 4 micron
A high-performance spherical silica carrier with a mean particle size of 4μm provides an excellent pressure/separation performance ratio
Manufacturer: emd millipore  1502040001
 View more versions of this product
Catalog No. M1502040001
  • / Each
Description & Specifications
Specifications
Quantity 3/Pk.
Diameter (English) Inside 2mm
Material Stainless-steel
Sorbent Superspher 100 RP-18 sorbent
Type HPLC Cartridges
Length (Metric) 10m
Particle Size 4um