MilliporeSigma™ LiChroCART™ Column/LiChrospher™ WP 300 RP-18 Sorbent HPLC Cartridges, 5μm
HPLC guard column
$1,331.43 - $1,331.43
Specifications
Diameter (Metric) 4 mm
Material Stainless Steel
Type HPLC Cartridge
View More Specs
Products 1
Catalog Number Mfr. No. Length (Metric) Quantity Price Quantity  
Catalog Number Mfr. No. Length (Metric) Quantity Price Quantity  
M1501400001
View Documents
MilliporeSigma™
1501410001
4 mm 10/Pk.
Each for $1,331.43
Description
Description
Purospher™ RP-18 is designed for the separation of problematic basic compounds with simple neutral eluents. In addition Purospher™ RP-18 allows the separation of hydrophilic compounds with aqueous mobile phase. Purospher™ RP-18 is based upon a high-purity, metal free silica. The multi-step chemical modification and deactivation of the surface enables a peak-tailing free elution of basic and chelating compounds. Purospher™ RP-18 is not suitable for the separation of acidic compounds. The good chemical stability of Purospher™ RP-18 enables the use of mobile phase conditions at pH 8 for a long period of time.
Specifications
Specifications
4 mm
Stainless Steel
HPLC Cartridge
SDS