MilliporeSigma™ LiChroCART™ Column/LiChrospher™ Si 60 Sorbent HPLC Cartridges, 5μm
HPLC cartridge
$1,461.62 - $2,092.15
Specifications
Diameter (Metric) 4 mm
Material Stainless Steel
View More Specs
Products 2
Catalog Number Mfr. No. Length (Metric) Quantity Type Price Quantity  
Catalog Number Mfr. No. Length (Metric) Quantity Type Price Quantity  
M1509280001
View Documents Promotion Details
MilliporeSigma™
1509280001
25 mm 3/Pk. HPLC Cartridge
Each for $2,092.15
M1509550001
View Documents Promotion Details
MilliporeSigma™
1509550001
4 mm 10/Pk. HPLC Cartridge
Each for $1,461.62
Description
Description
LiChrospher™ Si 60 is a versatile HPLC spherical silica sorbent with a pore diameter of 60A and a particle size of 5μm for normal phase chromatography: This sorbent is packed into a LiChroCART™ HPLC cartridge. The connection to the HPLC system is being accomplished by use of the manu-CART™ NT cartrige holder (1.51486).
Specifications
Specifications
4 mm
Stainless Steel