Please login to your online account to display your discounted pricing
EMD Millipore LiChroCART™ Column/LiChrospher™ 100 RP-8 Sorbent HPLC Cartridges, 5μm
Manufacturer: emd millipore  1506340001
 View more versions of this product
Catalog No. M1506340001
  • / Each
Description & Specifications
Specifications
Type HPLC Cartridges
Quantity 1/Pk.
Diameter (English) Inside 4.6mm
Length (Metric) 100mm
Material Stainless Steel