MilliporeSigma™ Vacuum Tubing
Accessories for filter holders for sample preparation.
Supplier:  MilliporeSigma™ XX6700037
 View more versions of this product
Catalog No. XX6700037
Description
Description
Millipore has a broad selection of fittings, adapters, valves, gauges, and tubing that can be used in conjunction with Millipore filtration systems. With stated exceptions, material of construction is Series 300 stainless steel.Tri-Clamp (TC) fittings are referred to by convention as 1 1/2 in., 2 in., etc. 1 1/2 in. TC fittings connect to 1 in. or 1 1/2 in. O.D. tube sizes. 2 in. TC fittings connect to 2 in. O.D. tube size only.
Specifications
Specifications
Laboratory Filter Holders or Cyclones
10 ft.
305 cm
Latex