Please login to your online account to display your discounted pricing

EMD Millipore Labscale TFF System Reservoir


Easy to use for sample preparation

Manufacturer: emd millipore  XX42LSS11

 View more versions of this product

Catalog No. XX42LSS11

  • / Each
 

Description & Specifications

Specifications

Height (Metric) 45.7cm
Width (English) 11 in.
Dimensions (L x W x H) 8.5 x 11 x 18 in. (21.6 x 27.9 x 45.7cm)
Height (English) 18 in.
Length (Metric) 21.6cm
Max. Inlet Pressure 60psi
Width (Metric) 27.9cm
For Use With (Application) Concentrate, desalt and exchange buffers (typical starting volumes, 50 to 1000mL), provides necessary controls for process development and scale-up work
Product Type Labscale Kits System, 115V/60Hz
Volume (Metric) 15mL
Includes Graduated 500mL acrylic reservoir with retentate back pressure valve, feed and retentate pressure indicators, stirrer assembly, diaphragm pump assembly with tubing and power supply
Length (English) 8.5 in.
Quantity 1 Kit