MilliporeSigma™ Inline Filter Holder
Supports inline production of ultraclean liquids or gasses by pressure prefiltration
Manufacturer:  MilliporeSigma™ XX4304700
Catalog No. XX4304700
Description
Description
  • Polypropylene filter material
  • Filter holder constructed with three nylon hand knobs for filter change without disturbing attached plumbing
  • Stepped hose connector at both ends (0.6cm [1/4 in.] NPTF fittings) with connector sized for 6 to 9.5mm (0.2 to 0.4 in.) I.D. flexible tubing
  • Holds 42mm (1.7 in.) thick-depth prefilters or 47mm (1.9 in.) membrane prefilters
  • Filter Diameter: 47mm (1.9 in.)
Specifications
Specifications
47 mm
Inline Filter Holder
Polypropylene