Please login to your online account to display your discounted pricing

EMD Millipore Immobilon™ Western Chemiluminescent HRP Substrate (ECL)


Provides high sensitivity over a broad detection range at an economical price

Manufacturer: emd millipore  WBKLS0500

 View more versions of this product

Catalog No. WBKLS0500

  • / Each
 

Description & Specifications

Specifications

Sufficient For 5000cm2 Membrane
Substrate Type Western Chemiluminescent HRP
Substrate Quantity 2 x 250mL
Shelf Life 9 Months
Storage Requirements 2-8°C
For Use With (Application) Western blotting, dot or slot blotting
Includes 250mL luminol, 250mL peroxide solution