Please login to your online account to display your discounted pricing

EMD Millipore Immobilon™ Western Chemiluminescent HRP Substrate (ECL)


Provides high sensitivity over a broad detection range at an economical price

Manufacturer: emd millipore  WBKLS0100

 View more versions of this product

Catalog No. WBKLS0100

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Western blotting, dot or slot blotting
Sufficient For 1000cm2 Membrane
Includes 50mL luminol, 50mL peroxide solution
Substrate Quantity 2 x 50mL
Storage Requirements 2-8°C
Shelf Life 9 Months
Substrate Type Western Chemiluminescent HRP

  • Combines high sensitivity and low background
  • Allows detection of high- and low-abundance proteins in the mid-femtogram range
  • Lowers antibody consumption by two- to five-fold compared to less sensitive reagents
  • Compatible with PVDF and nitrocellulose membranes as well as all commonly used buffers and blocking reagents

Accessories

9 Months