missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Immobilon™-PSQ PVDF Membranes

For Western blots and protein sequencing

Manufacturer:  MilliporeSigma™ ISEQ15150

 View more versions of this product

Catalog No. ISEQ15150



Description

Description

Immobilon-PSQ Membrane:
  • 0.2 μm pore size and a large internal structure
  • Higher protein adsorption and sequencing yields than other membranes
  • Recommended for blotting proteins less than 20 kDa
  • Prevents blow-through of low molecular weight proteins
  • Specifications

    Specifications

    15 cm
    For sequencing low-abundance or low-molecular weight (<20,000 Daltons) proteins
    Immobilon-PSQ
    15 x 15 cm
    15 cm
    Sheet
    0.2 μm
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions