missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Hydrophobic 6mm Edge Membranes

For microbiological and sterility testing of antibiotics and bacteriostatic drugs, as well as for filling machine operations

Manufacturer:  MilliporeSigma™ GSEP0470W

 View more versions of this product

Catalog No. GSEP0470W



Description

Description

  • Nonwetting edge prevents solution intrusion under holder sealing rings
  • Hydrophobic edge membranes are standard MF-Type (mixed cellulose ester) filters
  • 47mm diameter

Specifications

Specifications

White
4.7 cm
Plain
Mixed Cellulose Ester
Non-sterile
1.85 in.
Microbiological and sterility testing of antibiotics and bacteriostatic drugs, as well as for filling machine operations, filling machine operations, sterility testing
Sterile pack: 10 resealable envelopes containing 10 filters and 10 pads
0.22 μm
SDS
Documents
Product Certifications