MilliporeSigma™ Hydrophobic Edge Membranes
For microbiological and sterility testing of antibiotics and bacteriostatic drugs, as well as for filling machine operations
Manufacturer:  MilliporeSigma™ HAEP047S0
 View more versions of this product
Catalog No. HAEP047S0
Description
Description
Hydrophobic edge membranes are standard MF-Type filters with edges. The non-wetting edge prevents drug intrusion under holder sealing rim
Specifications
Specifications
White
0.45 μm
Clinical; Cosmetics; Environmental; Food and Beverage; Industrial QC; Pharmaceutical
Sterile-packed (10 resealable envelopes containing 10 filters and 10 pads, sterile)
Hydrophobic Edge Membrane
4.7 cm
Mixed Cellulose Esters (MCE)
Sterile
Hydrophobic Edge
Plain
1.85 in.
4.7 cm