EMD Millipore™ Hemacolor™ Staining Kit for Microscopy
staining set for microscopy
$247.17 - $413.06
Specifications
For Use With (Application) Rapid staining of blood smear
Quantity 1 set
Type Hemacolor™ Staining Kit
View More Specs
Products 2
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
M1116610001 View Documents EMD Millipore™
1116610001
Each for $413.06
M1116740001 View Documents EMD Millipore™
1116740001
Each for $247.17
Description
Description
The Hemacolor™ staining kit combines the brilliance and reproducibility of the Pappenheim staining with the time-advantage of a rapid stain. The Hemacolor™ staining kit contains a fixing solution, one red (eosin) and one blue (azur) staining solution and phosphate buffer tablets pH 7.2 acc. to Weise. The staining solutions are separately applied, which results in a high degree of stability. The conventionally appearing precipitates in the hematological staining solutions, could be avoided in this kit. In addition to blood and bone marrow smears, Hemacolor™ can be used in clinical cytology, e.g. for staining urine, sputum, FNAB, sperm, effusions and lavages. A kit contains safe 100-ml or 500-ml PE-bottles and six buffer tablets; sufficient for staining 250 and up to 1000 slides. The Hemacolor™ staining kit is certified and registered as CE / IVD product for applications in diagnostics.
Specifications
Specifications
Rapid staining of blood smear
1 set
Hemacolor™ Staining Kit