EMD Millipore Hemacolor™ Staining Kit for Microscopy
staining set for microscopy
Manufacturer:  EMD Millipore 1116740001
 View more versions of this product
Catalog No. M1116740001
Specifications
Description
Kit, Staining; EMD Millipore; Hemacolor; Rapid staining of blood smear; Includes Solution 1 fixing solution (100mL), Solution 2 color reagent red (100mL), Solution 3 color reagent blue (100mL); 3 Buffer tablets pH 7.2 according to Weise
The Hemacolor® staining kit combines the brilliance and reproducibility of the Pappenheim staining with the time-advantage of a rapid stain. The Hemacolor® staining kit contains a fixing solution, one red (eosin) and one blue (azur) staining solution and phosphate buffer tablets pH 7.2 acc. to Weise. The staining solutions are separately applied, which results in a high degree of stability. The conventionally appearing precipitates in the hematological staining solutions, could be avoided in this kit. In addition to blood and bone marrow smears, Hemacolor® can be used in clinical cytology, e.g. for staining urine, sputum, FNAB, sperm, effusions and lavages. A kit contains safe 100-ml or 500-ml PE-bottles and six buffer tablets; sufficient for staining 250 and up to 1000 slides. The Hemacolor® staining kit is certified and registered as CE / IVD product for applications in diagnostics.
Description & Specifications
Specifications
1 set
Hemacolor™ Staining Kit
Rapid staining of blood smear
Solution 1 (100mL), Solution 2 (100mL), Solution 3 (100mL); 3 Buffer tablets pH 7.2