Please login to your online account to display your discounted pricing

EMD Millipore Harleco™ Wright Stain Solutions

For use with the MIDAS™ III Automated Stainer

$157.54 - $762.97

Specifications

Boiling Point 64.5°C
Type Wright Stain Solution
Format Liquid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

36-102-0552

 
emd millipore
74075
1L Each for $157.54

36-102-0495

 
emd millipore
74085
4L Each for $322.06

23750077

 
emd millipore
74086
10L Each for $762.97
Description & Specifications

Specifications

Boiling Point 64.5°C
Type Wright Stain Solution
Format Liquid

  • Polychromic stain for peripheral blood and bone marrow smears
  • Used for red blood cell morphology and white blood cell differentiation

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.