Please login to your online account to display your discounted pricing

EMD Millipore HARLECO™ Wright's Stain

For staining blood cells

$256.77 - $740.20

Specifications

Format Bottle
For Use With (Application) For staining Blood cells
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

23749965

 
emd millipore
37616
100g Each for $740.20

23749964

 
emd millipore
37612
25g Each for $256.77
Description & Specifications

Specifications

Format Bottle
For Use With (Application) For staining Blood cells

  • Hematology stain mixture
  • Grade: Certified biological stain