Please login to your online account to display your discounted pricing
EMD Millipore Harleco™ Histology/Papanicolaou Stains: OG-6 Papanicolaou Stain
Harleco™ Histology/Papanicolaou Stains
$209.31 - $667.06
Specifications
Type OG-6 Papanicolaou Stain
For Use With (Application) Cytoplasmic counterstain for pap staining technique
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    
36-102-0551
emd millipore
7052X75
1000mL Each for $209.31
36-102-0531
emd millipore
7052X85
4000mL Each for $274.80
23750062
emd millipore
7052X86
10000mL Each for $667.06
Description & Specifications
Specifications
Type OG-6 Papanicolaou Stain
For Use With (Application) Cytoplasmic counterstain for pap staining technique
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.