Please login to your online account to display your discounted pricing

EMD Millipore HARLECO™ Fuchsin Carbol Solution

For acid-fast staining of micro-organisms

$329.09 - $552.74

Specifications

Format Bottle
For Use With (Application) Acid fast staining of microorganisms
Quantity 1L
Stain Type Carbolfuchsin
View More Specs
Products
Catalog Number Mfr. No. Type Price Quantity    

23-749979

 
emd millipore
6033X75
Kinyoun Stain Each for $552.74

23749981

 
emd millipore
6051X75
Ziehl Neelsen Formula Each for $329.09
Description & Specifications

Specifications

Format Bottle
For Use With (Application) Acid fast staining of microorganisms
Quantity 1L
Stain Type Carbolfuchsin

  • Stain for acid-fast mycobacteria
  • Ziehl-Neelsen formula/Kinyoun stain mixture
  • Packaged in poly bottle