Please login to your online account to display your discounted pricing

EMD Millipore Harleco™ Azure B Giemsa Stain Solution

For use with the MIDAS™ III Automated Stainer

$253.74 - $253.74

Specifications

Format Liquid
View More Specs
Products
Catalog Number Mfr. No. Quantity Type Price Quantity    

23749986

 
emd millipore
62075
1L Giemsa Stain Solution, Modified Azure Blend Each for $253.74
Description & Specifications

Specifications

Format Liquid

  • Polychromatic stain
  • Used as stand alone stain or in combination with Wright Stain for staining of blood parasites such as malaria and rickettsia

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.