missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Glass Fiber Filter without Binder

hydrophilic Glass Fiber without Binder Resin filter

$38.00 - $90.20

Specifications

Autoclavable Yes
Color White
For Use With (Application) Used for contamination analysis
Material Borosilicate Microfiber Glass
Filter Type Depth filter
View More Specs

Products 3
Catalog Number Mfr. No. Diameter (Metric) Outside Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Outside Price Quantity  
APFB03700 View Documents MilliporeSigma™
APFB03700
3.7cm Each for $90.20
 
AP4007000 View Documents MilliporeSigma™
AP4007000
7cm Pack of 100 Each for $84.10
 
AP4002500 View Documents MilliporeSigma™
AP4002500
2.5cm Pack of 100 Each for $38.00
 
Description

Description

hydrophilic Glass Fiber filter with a 0.7μm pore size is a recommended prefilter for 0.9 to 8um filters, especially for heavily contaminated liquids. Can be sterilized by EO, gamma and autoclave.
Specifications

Specifications

Yes
Used for contamination analysis
Depth filter
Plain
White
Borosilicate Microfiber Glass
Sterile
Hydrophilic