Please login to your online account to display your discounted pricing

EMD Millipore Fuchsin, HARLECO™


Acid Magenta; certified biological stain

Manufacturer: emd millipore  21816

 View more versions of this product

Catalog No. M21816

  • / Each
 

Description & Specifications

Specifications

Type Fuchsin Acid
Quantity 100g
Format Powder
For Use With (Application) Plasma or collagen fiber staining
Color Green