Please login to your online account to display your discounted pricing

EMD Millipore Fuchsin, HARLECO™


Acid Magenta; certified biological stain

Manufacturer: emd millipore  21812

 View more versions of this product

Catalog No. M21812

  • / Each
 

Description & Specifications

Specifications

Color Green
For Use With (Application) Plasma or collagen fiber staining
Format Powder
Type Fuchsin Acid
Quantity 25g