missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Fluoropore™ PTFE Membrane Filters

Hydrophobic membrane with laminated backing for easier handling

Manufacturer:  MilliporeSigma™ FSLW04700

 View more versions of this product

Catalog No. FSLW04700



Description

Description

  • Made from polytetrafluoroethylene (PTFE) bonded to high-density polyethylene
  • Wetting with methanol or another low-surface-tension fluid renders the membrane hydrophilic
  • Mean thickness varies from 145 to 200μm, depending upon filter type
  • Provides broad chemical compatibility
  • Autoclavable at 121°C (250°F)

Applications:

Clarification of acids, bases and solvents; clarification of alcohol solutions; UV spectroscopy.

Specifications

Specifications

Yes
PTFE
White
0.7psi
20.0L/min./cm sq.
3.0μm
47mm
100/pk.
Clarification of acids, bases and solvents; clarification of alcohol solutions; UV spectroscopy.
Documents