missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Fluoropore™ Membrane Filters, 0.5 μm

Hydrophobic membrane with laminated backing for easier handling

Supplier:  MilliporeSigma™ FHLP02500

 View more versions of this product

Catalog No. FHLP02500



Description

Description

Our Fluoropore™ membrane is a hydrophobic, polytetrafluoroethylene (PTFE) polymer membrane bonded to a high density polyethylene support to improve the handling characteristics of the filter for normal applications. Fluoropore™ membranes provide broad chemical compatibility, high flow rates and consistency. (Please note that catalogue numbers containing in.FHUP in. are unlaminated, or produced without a backing.)Applications: Fluoropore™ Membrane: Clarifying Acids, Bases and Solvents; Air Monitoring; Filtering and Venting Gases; UV Spectroscopy
Specifications

Specifications

Not Autoclavable
White
PTFE
Non-sterile
Clarification of Acids, Bases and Solvents; Clarification of Alcohol Solutions; UV Spectroscopy.
Hydrophobic
100/Pk.
Hydrophobic
25 mm
7 psi
Screen filter
0.45 μm
60 mL/min x cm2
8.0 L/min/cm2
100Pack
Filter Membranes
25 mm
SDS
Documents

Documents

Accessories
Promotions

Promotions