missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore Fixed Speed Peristaltic Pump: Replacement Parts

Provides constant flow of 80-100mL/min.

$309.90 - $812.39

Specifications

For Use With (Equipment) 15V/230V Fixed speed peristaltic pumps
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Tube Type Price Quantity  
Catalog Number Mfr. No. Tube Type Price Quantity  
XX8000024 EMD Millipore
XX8000024
Silicone tubing Each for $309.90
XX80EL004 EMD Millipore
XX80EL004
Pump Head Pack for $812.39
Specifications

Description

  • Polysulfone and steel pump head; stainless-steel chassis; painted steel case
  • Silicone tubing withstands 1.4 bar (20psi) pressure when operated for up to 20 hours at 500rpm
  • Silicone tubing: I.D.: 3/16 in., Length: 7.6m

Description & Specifications

Specifications

15V/230V Fixed speed peristaltic pumps