missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Accessories for YT30090HW ZHE Hazardous Waste System
Accessories for filter holders for sample preparation.
Supplier: MilliporeSigma™ XX3002567
Description
Accessories for filter holders for sample preparation.Specifications
NPTF to M | |
Laboratory Filter Holders or Cyclones | |
0.125 in. |
Water Purification System | |
Nickel/Chrome plated | |
0.32 cm |