Please login to your online account to display your discounted pricing
EMD Millipore Filter Holder Vacuum Manifolds
Provide vacuum support for simultaneous filtration of either three or six test samples
Manufacturer: emd millipore  XX2604735
 View more versions of this product
Catalog No. XX2604735
  • / Each
Description & Specifications
Specifications
Description Manifold
Material PVC
For Use With (Equipment) Vacuum Filter Holder