MilliporeSigma™ Filter Forceps Edge
Edge
Handle membrane filters without damage
Supplier:  MilliporeSigma™ XX6200006P
Catalog No. XX6200006P
Edge
Description
Description
  • Made from highly polished stainless steel with beveled, unserrated tips
  • Sterilize by autoclaving or flaming
  • Pack of 3; contains blue, pink and green
Specifications
Specifications
Beveled, Unserrated
Stainless Steel
Forceps, Filter
3/Pack