Please login to your online account to display your discounted pricing

EMD Millipore Fast-Trap™ Adenovirus and Lentivirus Purification and Concentration Kits


Quick, easy membrane-based method for laboratory-scale virus purification

Manufacturer: emd millipore  FTAV00003

 View more versions of this product

 Alert

These kits permit quick purification of infectious agents which should be handled under BSL2 safety precautions. Wear hand, eye, face, and body personal protective equipment (PPE) when using this kit. All virus purification steps (except water bath incubation and centrifugation) should be conducted in a biosafety cabinet.

 Disclaimers

For laboratory use only; not for diagnostic or in vivo use.

Catalog No. FTAV00003

  • / Each
 

Description & Specifications

Specifications

No. of Preps 3.0
Includes Three fast trap virus filter units, three steriflip HV filter units, 20mL 10X Binding Buffer, 100mL Equilibration Buffer, 200mL Wash Buffer, 21mL Elution Buffer, three Amicon ultra 4 filter units, test tube stand
Test Time <2 hr.
For Use With (Application) Adenovirus purification and concentration
Yield 1 x 1013 virus particles
Storage Requirements Room Temperature
Product Type Fast trap adenovirus purification and concentration kit