missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Fast-Trap™ Adenovirus and Lentivirus Purification and Concentration Kits

Quick, easy membrane-based method for laboratory-scale virus purification

$827.68 - $829.04

Specifications

Product Type Fast trap adenovirus purification and concentration kit
For Use With (Application) Adenovirus purification and concentration
Includes Three fast trap virus filter units, three steriflip HV filter units, 20mL 10X Binding Buffer, 100mL Equilibration Buffer, 200mL Wash Buffer, 21mL Elution Buffer, three Amicon ultra 4 filter units, test tube stand
No. of Preps 3.0
Storage Requirements Room Temperature
View More Specs

Alert:

These kits permit quick purification of infectious agents which should be handled under BSL2 safety precautions. Wear hand, eye, face, and body personal protective equipment (PPE) when using this kit. All virus purification steps (except water bath incubation and centrifugation) should be conducted in a biosafety cabinet.


Products 2
Catalog Number Mfr. No. Yield Price Quantity  
Catalog Number Mfr. No. Yield Price Quantity  
FTAV00003 View Documents MilliporeSigma™
FTAV00003
1 x 1013 virus particles Each for $829.04
 
FTLV00003 View Documents MilliporeSigma™
FTLV00003
2 x 108 virus particles Each for $827.68
 
Description

Description

Fast-Trap Purification

  • Saves time—protocol can be completed in less than two hours
  • High yield—recover up to 70% of virus particles
  • Dependable—simple protocol ensures quality results
  • Easy—Steriflip™ filters eliminate messy purification steps and expensive equipment
  • Safe—use a closed vacuum device to eliminate potential spills

Specifications

Specifications

Fast trap adenovirus purification and concentration kit
Three fast trap virus filter units, three steriflip HV filter units, 20mL 10X Binding Buffer, 100mL Equilibration Buffer, 200mL Wash Buffer, 21mL Elution Buffer, three Amicon ultra 4 filter units, test tube stand
Room Temperature
Adenovirus purification and concentration
3.0
<2 hr.

For laboratory use only; not for diagnostic or in vivo use.