missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore™ EZ-Fit™ Filtration Unit

For bioburden testing of liquid samples like water, process samples, or final products

Manufacturer:  EMD Millipore™ EFHAB250B

 View more versions of this product

Catalog No. EFHAB250B



Description

Description

  • Designed for optimizing and securing laboratory workflow to provide time-saving and reliable microbiological results
  • Comprise of 100mL or 250mL funnels
  • Black or white gridded membranes
  • After filtration of sample, membrane can be transferred and cultured on agar plate
  • Sterilized and disposable
  • Units are either individually bagged or packaged in bulk
Conform to international standards (EP/USP) and comply with water testing regulations.
Specifications

Specifications

Membrane: Black
Mixed cellulose esters
Sterile
0.45μm
48
Bulk packaging
48/Pk.
250mL
250mL
Documents