MilliporeSigma™ EXtrelut NT Prepacked Column: Filters
Spare filters for EXtrelut™ NT prepacked glass columns
Supplier: MilliporeSigma™ 1145670001
Description
Extrelut™ NT simplifies liquid-liquid extraction in its most effective form by using a chemically inert, wide-pore and highly pure diatomaceous earth based solid phase. Depending on the sample volume three types of columns are available: Extrelut™ NT1 can take up a maximum of one ml, Extrelut™ NT3 threemL and Extrelut™ NT20 up to twentymL of sample.Specifications
Replacement Filter | |
50 Units | |
EXtrelut NT 20 HDPE Prepacked Columns |