missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Express™ PLUS Membrane Filters
Plain white polyethersulfone Express™ PLUS Membrane Filters provide ultrafast filtration for a variety of applications.
Supplier: MilliporeSigma™ GPWP14250
Description
Plain white polyethersulfone membranes provide ultrafast filtration for a variety of applications
The Millipore Express™ PLUS membrane provides ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This high throughput, low protein-binding membrane is also used in many of our ready-to-use sterile filtration devices.
Features and Benefit:- Ultra fast filtration and high throughput shorten process time
- Can be sterilized by autoclave, ethylene oxide, or gamma irradiation for sterile filtration applications
- Applications:Sterile Filtration, Buffer Filtration, Tissue Culture Media Filtration
Specifications
| Not Autoclavable | |
| Reusable | |
| Disc | |
| 0.22 μm | |
| 15 (at 690 Mbar (10 psi)) | |
| Hydrophilic | |
| Hydrophilic | |
| 142 mm |
| White | |
| Screen Filter | |
| PES | |
| Non-sterile | |
| Provide ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This membrane is also used in many ready-to-use sterile filtration devices. | |
| Filter Membranes | |
| 5.6 in. |