missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Express™ PLUS Membrane Filters
Plain white polyethersulfone membranes provide ultrafast filtration for a variety of applications
Manufacturer: MilliporeSigma™ HPWP14250
Description
Plain white polyethersulfone membranes provide ultrafast filtration for a variety of applications
- Hydrophilic, high-throughput, low-protein-binding
- Thickness: ≥150μm and ≤200μm
- Sterilization: by autoclave (121°C at 1 bar), EO or gamma
- Bacterial endotoxins: 0.5 EU/mL
- Gravimetric extractables: <1.5%
- Flowrate: 40mL/min. cm2 at 2 bar, 25°C
Applications:
Provide ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This membrane is also used in many ready-to-use sterile filtration devices.
Specifications
Autoclavable | |
142 mm | |
Hydrophilic | |
0.45 μm |
White | |
Provide ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This membrane is also used in many ready-to-use sterile filtration devices. | |
PES |