MilliporeSigma™ Express™ PLUS Membrane Filters
Plain white polyethersulfone membranes provide ultrafast filtration for a variety of applications
Supplier:  MilliporeSigma™ GPWP02500
 View more versions of this product
Catalog No. GPWP02500
Description
Description
Plain white polyethersulfone membranes provide ultrafast filtration for a variety of applications
The Millipore Express™ PLUS membrane provides ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This high throughput, low protein-binding membrane is also used in many of our ready-to-use sterile filtration devices.Features & Benefit:•Ultra fast filtration and high throughput shorten process time •Can be sterilized by autoclave, ethylene oxide, or gamma irradiation for sterile filtration applications Applications:Sterile Filtration, Buffer Filtration, Tissue Culture Media Filtration
Specifications
Specifications
Not Autoclavable
Screen Filter
0.22 μm
15 (at 690 Mbar (10 psi))
Hydrophilic
Hydrophilic
25 mm
White
PES
Non-sterile
Provide ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This membrane is also used in many ready-to-use sterile filtration devices.
Filter Membranes
25 mm
SDS
Documents
Documents
Accessories