Please login to your online account to display your discounted pricing

EMD Millipore™ EmbryoMax™ Modified M16 Medium (1X), Powdered, w/o phenol red


Manufacturer: emd millipore corporation  MR010P5F

 View more versions of this product

Catalog No. MR010P5F

  • / Each
 

Description & Specifications

Specifications

Storage Requirements 2- 8°C for 6 - 12 months
For Use With (Application) Stem cell culture and embryo culture
Quantity 50mL