MilliporeSigma™ EmbryoMax™ M2 Medium (1x), Liquid, with Phenol Red and Hyaluronidase
Optimized and validated for Embryo Culture.
Supplier: MilliporeSigma™ MR051F
Description
Dry Powdered form offers easy and quick preparation, and the freshest nutrients to support the development of embryos in culture. Just add the sterile diluent (included in the kit) to the powder, mix gently, and filter to prepare FRESH medium with the correct pH and osmolarity.Specifications
-20°C | |
EmbryoMax™ M2 Medium (1X) with Phenol Red and Hyaluronidase | |
Stem Cell Culture, Embryonic Cell Culture | |
10 mL |