Please login to your online account to display your discounted pricing

EMD Millipore™ EmbryoMax™ M2 Medium (1X) Powdered With Phenol Red


Manufacturer: emd millipore corporation  MR015P5D

 View more versions of this product

Catalog No. MR015P5D

  • / Each
 

Description & Specifications

Specifications

Quantity 250mL
Storage Requirements 2- 8°C
For Use With (Application) Stem cell culture and embryo culture