Please login to your online account to display your discounted pricing

EMD Millipore™ EmbryoMax™ M2 Medium (1x)


Manufacturer: emd millipore corporation  MR015PD

 View more versions of this product

Catalog No. MR015PD

  • / Each
 

Description & Specifications

Specifications

Quantity 50mL
Storage Requirements 2-8°C
For Use With (Application) Embryo and stem cell culture