EMD Millipore™ EmbryoMax™ KSOM Embryo Culture (1x) w/o Phenol Red
Optimized and validated for Embryo Culture.
$56.49 - $238.27
Specifications
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MR020P5D Emd Millipore Corporation
MR020P5D
250mL Each for $238.27
MR020P5F Emd Millipore Corporation
MR020P5F
50mL Each for $88.76
MR020PD Emd Millipore Corporation
MR020PD
50mL Each for $56.49
Specifications
Description
We now offer our most popular embryo culture media in a dry powder format. Each has been formulated for optimal, consistent performance. This new dry powdered format offers greatly increased shelf-life, easy and quick preparation, and the freshest nutrients to support the development of your embryos in culture. Just add the sterile diluent (included in the kit) to the powder, mix gently, and filter to prepare FRESH medium with the correct pH and osmolarity.
Description & Specifications
Specifications
0.0