Please login to your online account to display your discounted pricing

EMD Millipore™ EmbryoMax™ 1X Dulbecco's Phosphate Buffered Saline w/o Ca&superscript_plus;&superscript_plus; and Mg&superscript_plus;&superscript_plus;


Manufacturer: emd millipore corporation  BSS1006B

 View more versions of this product

Catalog No. BSS1006B

  • / Each
 

Description & Specifications

Specifications

Quantity 500mL
Storage Requirements 15-30°C
For Use With (Application) Stem cell culture