Please login to your online account to display your discounted pricing

EMD Millipore Elix™ Advantage Water Purification System


With the Millipore Elix Advantage Water Purification System, you can produce pure water that meets the highest standards, together with an ergonomic state-of-the art water dispensing system. Peace of mind and ease of use—all in one!

Manufacturer: emd millipore  ZRXV015US

 View more versions of this product

Catalog No. ZRXV015US

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Width (English) 13.7 in.
For Use With (Application) Reagent and buffer preparation, microbiology media preparation, feed for laboratory equipment
Product Type Water electrode ionization system
Feedwater Source Potable tap water
Height (English) 19.6 in.
Hertz 50/60Hz
Model Elix™ Advantage 15
Modules Elix™
Total Organic Carbon Less than30
Voltage 100/230V
Width (Metric) 35cm
Certifications/Compliance ASTM D1193, ISO 3696
For Use With (Equipment) E-POD unit
Daily Output 350L
Depth (English) 18.8 in.
Electrical Requirements 100/230V 50/60Hz
Range (Conductivity) 0.067 to 0.1
Depth (Metric) 48cm
Flow Rate 15
Height (Metric) 50cm
Conductivity 0.067 to 0.1μS/cm
Resistivity 5 to 15
Product Water Type Type 2

Elix electrodeionization technology
Combining Millipore Elix electrodeionization technology with the most advanced purification technologies (Progard™ pretreatment, advanced reverse osmosis, 254nm UV lamp), the Elix Advantage system uses potable tap water as feed. The pure water produced by the system meets the most rigorous standards of regulatory bodies around the globe.

Ergonomic E-POD™ units for flexible water delivery

  • Up to three E-POD units per system can be installed at convenient locations in your lab, letting you benefit from efficient and reliable pressurized pure water delivery, with manual or volumetric dispensing.
Save time and money; benefit from lower running costs and reduced maintenance
  • Produce up to 350L per day
  • No polishing pack needed for pure water production
  • No need for an anti-scaling cartridge upstream of the Elix module
  • No extra softeners required
Peace of mind, ease of use
  • Consistent water quality, with resistivity measurements displayed on the main unit's graphic display
  • Continuous electrical recharging of exchange resins elilminates variations due to exhaustion of resins common to chemically regenerated units
  • High resistance to scaling ensures high Elix module reliability
  • All-in-one pretreatment pack
  • UV lamp maintains low bacteria levels
  • Unit performs self-maintenance of reverse osmosis membrane
  • Provides pure water for compliance with industry standards
Planet-friendly, clean technology
  • Uses electricity rather than harsh chemicals to regenerate resins and remove ions
  • Minimal use of disposable purification cartridges limits waste
  • Low electricity consumption
  • High water recovery minimizes tap water waste
  • Data on system operation and performance appear on the main unit's display
  •  
Information at your fingertips
  • Daily output data appear on the E-POD screen: water quality, system status, tank level, maintenance messages, water dispensing information
Adapability to meet your needs
  • Compact unit can be installed on the wall or under a sink, saving valuable bench space
  • Modular design adapts to changing demands in your lab: up to three E-POD units can be added
  • Choice of two dispensing flowrates and manual or automatic dispensing modes

 

Reagent and buffer preparation, microbiology media preparation, histology applications, purified water for pharmaceutical applications (according to United States, Japanese, and European Pharmacopeias), and feed for laboratory equipment (washing machines, clinical analyzers, stability chambers, humidifiers, autoclaves, and weatherometers)