missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Durapore™ PVDF Membrane Filters: Hydrophobic

Recommended when maximum recovery is important

Manufacturer:  MilliporeSigma™ GVHP04700

 View more versions of this product

Catalog No. GVHP04700



Description

Description

Recommended when maximum recovery is important

  • Mean thickness: 125μm
  • Typical refractive index: 1.42
  • Autoclavable
  • Hydrophilic form is made from modified polyvinylidene fluoride (PVDF); lower protein binding than nitrocellulose, nylon, or PTFE

Hydrophobic:

  • Clarification of air/gases; sterilizing filtration of gases

Specifications

Specifications

Yes
Polyvinylidene Fluoride (PVDF)
White
0.2μm
47mm
100/pk.
Documents