Please login to your online account to display your discounted pricing

EMD Millipore Durapore™ PVDF Membrane Filters: Hydrophilic: 5.0μ Pore Size


Recommended when maximum recovery is important

Manufacturer: emd millipore  SVLP01300

 View more versions of this product

Catalog No. SVLP01300

  • / Pack of 100 Each
 

Description & Specifications

Specifications

Material Polyvinylidene Fluoride (PVDF)
Diameter (Metric) Outer 13mm
Color White; No Grid
Quantity (No. per Case) 100/pk.
Autoclavable Yes
Bubble Point 3psi
Liquid Flow Rate 288.0mL/min./cm sq.
Wettability Hydrophilic

  • Mean thickness: 125μm
  • Typical refractive index: 1.42
  • Autoclavable
  • Hydrophilic form is made from modified polyvinylidene fluoride (PVDF); lower protein binding than nitrocellulose, nylon, or PTFE

Applications:

  • Clarification of air and other gases
  • Sterilizing filtration of protein solutions, tissue culture media, additives, antibiotics, and alcohols
  • Particulate removal; biological assays (receptor binding studies, TCA precipitates)